Learn More
Colistin sodium methanesulfonate, Thermo Scientific™
£117.99 - £476.02
Chemical Identifiers
| CAS | 8068-28-8 |
|---|---|
| Molecular Formula | C58H105N16Na5O28S5 |
| Molecular Weight (g/mol) | 1749.811 |
| MDL Number | MFCD00130824 |
| InChI Key | IQWHCHZFYPIVRV-KLWRPAJRSA-I |
| Synonym | Colimycin sodium methanesulfonate; Polymyxin E sodium methanesulfonate |
| PubChem CID | 131668505 |
| IUPAC Name | pentasodium;(6S)-N-[(2R)-1-[[(2R,3S)-3-hydroxy-1-[[(2S)-1-[[(3S,6S,9S,12S,15R,18S,21S)-3-[(1R)-1-hydroxyethyl]-12,15-bis(2-methylpropyl)-8,17,20-trioxido-2,5,11,14-tetraoxo-6,9,18-tris[2-(sulfomethylamino)ethyl]-1,4,7,10,13,16,19-heptazacyclotricosa-7,16, |
| SMILES | CCC(C)CCCCC(=NC(CCNCS(=O)(=O)O)C(=O)NC(C(C)O)C(=O)NC(CCNCS(=O)(=O)O)C(=NC1CCNC(=O)C(NC(=O)C(N=C(C(NC(=O)C(NC(=O)C(N=C(C(N=C1[O-])CCNCS(=O)(=O)O)[O-])CC(C)C)CC(C)C)CCNCS(=O)(=O)O)[O-])CCNCS(=O)(=O)O)C(C)O)[O-])[O-].[Na+].[Na+].[Na+].[Na+].[Na+] |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15418159
|
Thermo Scientific Alfa Aesar
J66698.03 |
1g |
£121.00 £117.99 / 1g Save £3.01 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15428159
|
Thermo Scientific Alfa Aesar
J66698.06 |
5g |
£486.00 £476.02 / 5g Save £9.98 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilityFreely soluble in water
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Chemical Identifiers
| 8068-28-8 | |
| 1749.811 | |
| IQWHCHZFYPIVRV-KLWRPAJRSA-I | |
| 131668505 | |
| CCC(C)CCCCC(=NC(CCNCS(=O)(=O)O)C(=O)NC(C(C)O)C(=O)NC(CCNCS(=O)(=O)O)C(=NC1CCNC(=O)C(NC(=O)C(N=C(C(NC(=O)C(NC(=O)C(N=C(C(N=C1[O-])CCNCS(=O)(=O)O)[O-])CC(C)C)CC(C)C)CCNCS(=O)(=O)O)[O-])CCNCS(=O)(=O)O)C(C)O)[O-])[O-].[Na+].[Na+].[Na+].[Na+].[Na+] |
| C58H105N16Na5O28S5 | |
| MFCD00130824 | |
| Colimycin sodium methanesulfonate; Polymyxin E sodium methanesulfonate | |
| pentasodium;(6S)-N-[(2R)-1-[[(2R,3S)-3-hydroxy-1-[[(2S)-1-[[(3S,6S,9S,12S,15R,18S,21S)-3-[(1R)-1-hydroxyethyl]-12,15-bis(2-methylpropyl)-8,17,20-trioxido-2,5,11,14-tetraoxo-6,9,18-tris[2-(sulfomethylamino)ethyl]-1,4,7,10,13,16,19-heptazacyclotricosa-7,16, |
Specifications
| 8068-28-8 | |
| C58H105N16Na5O28S5 | |
| 14,2479 | |
| Freely soluble in water | |
| CCC(C)CCCCC(=NC(CCNCS(=O)(=O)O)C(=O)NC(C(C)O)C(=O)NC(CCNCS(=O)(=O)O)C(=NC1CCNC(=O)C(NC(=O)C(N=C(C(NC(=O)C(NC(=O)C(N=C(C(N=C1[O-])CCNCS(=O)(=O)O)[O-])CC(C)C)CC(C)C)CCNCS(=O)(=O)O)[O-])CCNCS(=O)(=O)O)C(C)O)[O-])[O-].[Na+].[Na+].[Na+].[Na+].[Na+] | |
| 1749.811 | |
| 1759.89 |
| White to Yellow | |
| MFCD00130824 | |
| Colimycin sodium methanesulfonate; Polymyxin E sodium methanesulfonate | |
| IQWHCHZFYPIVRV-KLWRPAJRSA-I | |
| pentasodium;(6S)-N-[(2R)-1-[[(2R,3S)-3-hydroxy-1-[[(2S)-1-[[(3S,6S,9S,12S,15R,18S,21S)-3-[(1R)-1-hydroxyethyl]-12,15-bis(2-methylpropyl)-8,17,20-trioxido-2,5,11,14-tetraoxo-6,9,18-tris[2-(sulfomethylamino)ethyl]-1,4,7,10,13,16,19-heptazacyclotricosa-7,16, | |
| 131668505 | |
| Colistin sodium methanesulfonate |
Safety and Handling
missing translation for 'einecsNumber' : 232-516-9
missing translation for 'rtecsNumber' : GH1650000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.