Learn More
Colistin sodium methanesulfonate, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J66698.03
| Quantity | 1g |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilityFreely soluble in water
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Chemical Identifiers
| 8068-28-8 | |
| 1749.811 | |
| IQWHCHZFYPIVRV-KLWRPAJRSA-I | |
| 131668505 | |
| CCC(C)CCCCC(=NC(CCNCS(=O)(=O)O)C(=O)NC(C(C)O)C(=O)NC(CCNCS(=O)(=O)O)C(=NC1CCNC(=O)C(NC(=O)C(N=C(C(NC(=O)C(NC(=O)C(N=C(C(N=C1[O-])CCNCS(=O)(=O)O)[O-])CC(C)C)CC(C)C)CCNCS(=O)(=O)O)[O-])CCNCS(=O)(=O)O)C(C)O)[O-])[O-].[Na+].[Na+].[Na+].[Na+].[Na+] |
| C58H105N16Na5O28S5 | |
| MFCD00130824 | |
| Colimycin sodium methanesulfonate; Polymyxin E sodium methanesulfonate | |
| pentasodium;(6S)-N-[(2R)-1-[[(2R,3S)-3-hydroxy-1-[[(2S)-1-[[(3S,6S,9S,12S,15R,18S,21S)-3-[(1R)-1-hydroxyethyl]-12,15-bis(2-methylpropyl)-8,17,20-trioxido-2,5,11,14-tetraoxo-6,9,18-tris[2-(sulfomethylamino)ethyl]-1,4,7,10,13,16,19-heptazacyclotricosa-7,16, |
Specifications
| 8068-28-8 | |
| 1g | |
| MFCD00130824 | |
| Colimycin sodium methanesulfonate; Polymyxin E sodium methanesulfonate | |
| IQWHCHZFYPIVRV-KLWRPAJRSA-I | |
| pentasodium;(6S)-N-[(2R)-1-[[(2R,3S)-3-hydroxy-1-[[(2S)-1-[[(3S,6S,9S,12S,15R,18S,21S)-3-[(1R)-1-hydroxyethyl]-12,15-bis(2-methylpropyl)-8,17,20-trioxido-2,5,11,14-tetraoxo-6,9,18-tris[2-(sulfomethylamino)ethyl]-1,4,7,10,13,16,19-heptazacyclotricosa-7,16, | |
| 131668505 | |
| Colistin sodium methanesulfonate |
| White to Yellow | |
| C58H105N16Na5O28S5 | |
| 14,2479 | |
| Freely soluble in water | |
| CCC(C)CCCCC(=NC(CCNCS(=O)(=O)O)C(=O)NC(C(C)O)C(=O)NC(CCNCS(=O)(=O)O)C(=NC1CCNC(=O)C(NC(=O)C(N=C(C(NC(=O)C(NC(=O)C(N=C(C(N=C1[O-])CCNCS(=O)(=O)O)[O-])CC(C)C)CC(C)C)CCNCS(=O)(=O)O)[O-])CCNCS(=O)(=O)O)C(C)O)[O-])[O-].[Na+].[Na+].[Na+].[Na+].[Na+] | |
| 1749.811 | |
| 1759.89 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.