missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL11A1 Antibody (SR1191), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | COL11A1 |
|---|---|
| Clone | SR1191 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18658414
|
Novus Biologicals
NBP3-22381-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18614225
|
Novus Biologicals
NBP3-22381-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COL11A1 Monoclonal antibody specifically detects COL11A1 in Human samples. It is validated for Western BlotSpecifications
| COL11A1 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 1301 | |
| IgG | |
| Affinity purified |
| SR1191 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| alpha-1 polypeptide, COLL6, collagen, type XI, alpha 1 | |
| A synthesized peptide derived from human COL11A1 (Uniprot #: P12107) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title