missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL11A1 Antibody (SR1191), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22381-100ul
This item is not returnable.
View return policy
Description
COL11A1 Monoclonal antibody specifically detects COL11A1 in Human samples. It is validated for Western Blot
Specifications
| COL11A1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1301 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| SR1191 | |
| Western Blot 1:500-1:2000 | |
| alpha-1 polypeptide, COLL6, collagen, type XI, alpha 1 | |
| A synthesized peptide derived from human COL11A1 (Uniprot #: P12107) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction