Learn More
DWK Life Sciences DURAN™ Coil Distillate Condenser, with 2 standard ground joints, and 2 screw-on plastic hose connections
The coil condenser serves to distill a reaction mixture or to cool a condensate.
Brand: DWK Life Sciences 242537208
Code : A0
Additional Details : Weight : 0.27000kg
Description
Typical for the coil condenser is its spiral riser tube, which is located in a jacket and thus washed around by the cooling medium (contrary to the Dimroth cooler). Due to this condensate flow, the condensation performance is very low although the cooling section is very long. This type of condenser is therefore usually used as a downstream additional condenser and not as an independent distillation condenser. Due to the better drainage of condensate in the convoluted riser tube, the condenser must be installed in a vertical position.
- Usually used as a downstream additional cooler due to long cooling section
- Two removable plastic olives for inlet and outlet of the cooling medium
- Tight closure due to standard ground joint
Specifications
NS 29/32 | |
Condensate cooling; Product condenser for distillate separations; Distillation | |
1 Ea. | |
Borosilicate Glass (3.3), PBT, Polypropylene, Silicone |