missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cofilin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£168.00
Specifications
| Antigen | Cofilin |
|---|---|
| Dilution | Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Cofilin Polyclonal specifically detects Cofilin in Human, Mouse, Rat, Porcine, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Cofilin | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cytoskeleton Markers | |
| 10mM PBS (pH 7.4) with 15mM Sodium Azide | |
| 18 kDa phosphoprotein, CFL, cofilin 1 (non-muscle), Cofilin, non-muscle isoform, cofilin-1, p18 | |
| CFL1 | |
| IgG | |
| IgG purified | |
| Detects a band at approximately 19kDa. Reacts with muscle and non-muscle cofilin. |
| Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| P23528 | |
| 1072 | |
| Synthetic peptide corresponding to amino acids 154-166 of human cofilin with N-terminal cysteine added, conjugated to maleimide-activated keyhole limpet hemocyanin (KLH). The corresponding immunizing sequence is identical in pig and rat non-muscle cofilin and differs by three amino acids from that of human and chicken muscle cofilin. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title