missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cofilin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-1900-0.025ml
This item is not returnable.
View return policy
Description
Cofilin Polyclonal specifically detects Cofilin in Human, Mouse, Rat, Porcine, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Cofilin | |
| Polyclonal | |
| Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:1000 | |
| P23528 | |
| CFL1 | |
| Synthetic peptide corresponding to amino acids 154-166 of human cofilin with N-terminal cysteine added, conjugated to maleimide-activated keyhole limpet hemocyanin (KLH). The corresponding immunizing sequence is identical in pig and rat non-muscle cofilin and differs by three amino acids from that of human and chicken muscle cofilin. | |
| 0.025 mL | |
| Cytoskeleton Markers | |
| 1072 | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 10mM PBS (pH 7.4) with 15mM Sodium Azide | |
| 18 kDa phosphoprotein, CFL, cofilin 1 (non-muscle), Cofilin, non-muscle isoform, cofilin-1, p18 | |
| Rabbit | |
| IgG purified | |
| RUO | |
| Primary | |
| Detects a band at approximately 19kDa. Reacts with muscle and non-muscle cofilin. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction