Learn More
Bollé Safety Cobra ESP Safety Glasses
Description
- One product = two uses
- Upper protection
- Panoramic vision
- Non-slip bridge
- Comfortable, non-slip straight temples
- Temple or strap version with interchangeable foam
- Pivoting temples
Specifications
Specifications
| Certifications/Compliance | Frame: EN 166 BT CE; Lens marking: 5-1.4 1 BT CE; Chemical risk - Liquid droplets - 3 (Goggle, adapted spectacles with strap or face shield mandatory); Mechanical risk - Low energy impact - F; Mechanical risk- High speed particles at extreme temperatures |
| Lens Coating | Anti-fog, Scratch-resistant |
| Description | COBRA wrap around safety glasses |
| For Use With (Equipment) | Black |
| Includes | B-ESP filters; Scratch-resistant, anti-fog (ASAF) |
| No. per Pack | 1 Ea. |
| Weight (Metric) | 25 g |
| Lens Material | Panoramic vision |
| Frame Material | Wraparound, non-slip bridge |