missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor II/Thrombin Antibody (GMA-020), Novus Biologicals™
Mouse Monoclonal Antibody
£174.00 - £581.00
Specifications
| Antigen | Coagulation Factor II/Thrombin |
|---|---|
| Clone | GMA-020 |
| Dilution | Western Blot 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18153464
|
Novus Biologicals
NBP2-29813-0.025MG |
0.025 mg |
£174.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18183072
|
Novus Biologicals
NBP2-29813 |
0.1 mg |
£581.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Coagulation Factor II/Thrombin Monoclonal specifically detects Coagulation Factor II/Thrombin in Human samples. It is validated for Western Blot.Specifications
| Coagulation Factor II/Thrombin | |
| Western Blot 1:100-1:2000 | |
| Monoclonal | |
| Mouse | |
| Apoptosis, Breast Cancer, Cancer, GPCR, Signal Transduction | |
| No buffer with No Preservative | |
| Coagulation Factor II, coagulation factor II (thrombin) receptor-like 2, Coagulation factor II receptor-like 2, Coagulation factor II receptor-like 2 (protease-actovated receptor 3), PAR-3, PAR3proteinase-activated receptor 3, protease-activated receptor 3, proteinase-activated receptor-3, PT, serine protease, Thrombin receptor-like 2 | |
| F2 | |
| IgG1 | |
| Affinity Purified | |
| 31 kDa |
| GMA-020 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| P00734 | |
| 2147 | |
| purified human thrombin. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title