missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor II/Thrombin Antibody (GMA-020), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-29813-0.025MG
This item is not returnable.
View return policy
Description
Coagulation Factor II/Thrombin Monoclonal specifically detects Coagulation Factor II/Thrombin in Human samples. It is validated for Western Blot.
Specifications
| Coagulation Factor II/Thrombin | |
| Monoclonal | |
| Unconjugated | |
| No buffer with No Preservative | |
| Coagulation Factor II, coagulation factor II (thrombin) receptor-like 2, Coagulation factor II receptor-like 2, Coagulation factor II receptor-like 2 (protease-actovated receptor 3), PAR-3, PAR3proteinase-activated receptor 3, protease-activated receptor 3, proteinase-activated receptor-3, PT, serine protease, Thrombin receptor-like 2 | |
| Mouse | |
| 31 kDa | |
| 0.025 mg | |
| Apoptosis, Breast Cancer, Cancer, GPCR, Signal Transduction | |
| 2147 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| GMA-020 | |
| Western Blot 1:100-1:2000 | |
| P00734 | |
| F2 | |
| purified human thrombin. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction