Learn More
Fisherbrand™ CO2 Backup Systems for Isotemp Chest Ultra-Low Temperature Freezers
Product Code. 17868010 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer
Isotemp 3 cu. ft. Chest Ultra-Low Temperature Freezer
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17868010 Isotemp 3 cu. ft. Chest Ultra-Low Temperature Freezer Each
17808020 Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17868010
Ensure optimal security for critical samples by using Fisherbrand™ Isotemp CO2 Backup Systems for Ultra-Low Temperature Freezers.
  • Maintains temperature down to -67°C with CO2
TRUSTED_SUSTAINABILITY
Specifications
Description Factory Installed
Temperature Range -67°C
For Use With (Equipment) Isotemp 3 cu. ft. Chest Ultra-Low Temperature Freezer