Learn More
Co 101244 hydrochloride, Tocris Bioscience™
GluN2B-selective NMDA antagonist
£146.08 - £473.65
Chemical Identifiers
CAS | 193356-17-1 |
---|---|
Molecular Formula | C21H28ClNO3 |
Molecular Weight (g/mol) | 377.909 |
InChI Key | WFFZHKKSIDENAJ-UHFFFAOYSA-N |
Synonym | co 101244 hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl piperidin-4-ol hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl-4-piperidinol mono hydrochloride, d08nrh, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl-4-piperidinol monohydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl-4-piperidinol hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-hydroxy-4-4-methylbenzyl piperidine hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4hydroxy-4-4-methylbenzyl piperidine hydrochloride |
PubChem CID | 6918426 |
IUPAC Name | 1-[2-(4-hydroxyphenoxy)ethyl]-4-[(4-methylphenyl)methyl]piperidin-4-ol;hydrochloride |
SMILES | CC1=CC=C(C=C1)CC2(CCN(CC2)CCOC3=CC=C(C=C3)O)O.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18064172
|
Tocris Bioscience
2456/10 |
10 mg |
£146.08
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18034592
|
Tocris Bioscience
2456/50 |
50 mg |
£473.65
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Novel, potent and selective antagonist of GluN2B (formally NR2B)-containing NMDA receptors (IC50 values are 0.043, > 100 and > 100 μM for GluN1A/2B (NR1A/2B), GluN1A/2A (NR1A/2A) and GluN1A/2C (NR1A/2C) subunit combinations respectively). Displays neuroprotective effects in vivo and in vitro.Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
Chemical Identifiers
193356-17-1 | |
377.909 | |
co 101244 hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl piperidin-4-ol hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl-4-piperidinol mono hydrochloride, d08nrh, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl-4-piperidinol monohydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl-4-piperidinol hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-hydroxy-4-4-methylbenzyl piperidine hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4hydroxy-4-4-methylbenzyl piperidine hydrochloride | |
1-[2-(4-hydroxyphenoxy)ethyl]-4-[(4-methylphenyl)methyl]piperidin-4-ol;hydrochloride |
C21H28ClNO3 | |
WFFZHKKSIDENAJ-UHFFFAOYSA-N | |
6918426 | |
CC1=CC=C(C=C1)CC2(CCN(CC2)CCOC3=CC=C(C=C3)O)O.Cl |
Specifications
193356-17-1 | |
co 101244 hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl piperidin-4-ol hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl-4-piperidinol mono hydrochloride, d08nrh, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl-4-piperidinol monohydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl-4-piperidinol hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-hydroxy-4-4-methylbenzyl piperidine hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4hydroxy-4-4-methylbenzyl piperidine hydrochloride | |
CC1=CC=C(C=C1)CC2(CCN(CC2)CCOC3=CC=C(C=C3)O)O.Cl | |
377.909 | |
377.91 | |
Co 101244 hydrochloride |
C21H28ClNO3 | |
WFFZHKKSIDENAJ-UHFFFAOYSA-N | |
1-[2-(4-hydroxyphenoxy)ethyl]-4-[(4-methylphenyl)methyl]piperidin-4-ol;hydrochloride | |
6918426 | |
>99% |
Safety and Handling
Recommended Storage : Store at +4°C