missing translation for 'onlineSavingsMsg'
Learn More

Co 101244 hydrochloride, Tocris Bioscience™

Catalog Number 18064172
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
193356-17-1
C21H28ClNO3
377.909
WFFZHKKSIDENAJ-UHFFFAOYSA-N
co 101244 hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl piperidin-4-ol hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl-4-piperidinol mono hydrochloride, d08nrh, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl-4-piperidinol monohydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl-4-piperidinol hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-hydroxy-4-4-methylbenzyl piperidine hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4hydroxy-4-4-methylbenzyl piperidine hydrochloride
6918426
1-[2-(4-hydroxyphenoxy)ethyl]-4-[(4-methylphenyl)methyl]piperidin-4-ol;hydrochloride
CC1=CC=C(C=C1)CC2(CCN(CC2)CCOC3=CC=C(C=C3)O)O.Cl
This item is not returnable. View return policy
193356-17-1
C21H28ClNO3
377.909
WFFZHKKSIDENAJ-UHFFFAOYSA-N
co 101244 hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl piperidin-4-ol hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl-4-piperidinol mono hydrochloride, d08nrh, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl-4-piperidinol monohydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl-4-piperidinol hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-hydroxy-4-4-methylbenzyl piperidine hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4hydroxy-4-4-methylbenzyl piperidine hydrochloride
6918426
1-[2-(4-hydroxyphenoxy)ethyl]-4-[(4-methylphenyl)methyl]piperidin-4-ol;hydrochloride
CC1=CC=C(C=C1)CC2(CCN(CC2)CCOC3=CC=C(C=C3)O)O.Cl

GluN2B-selective NMDA antagonist

Novel, potent and selective antagonist of GluN2B (formally NR2B)-containing NMDA receptors (IC50 values are 0.043, > 100 and > 100 μM for GluN1A/2B (NR1A/2B), GluN1A/2A (NR1A/2A) and GluN1A/2C (NR1A/2C) subunit combinations respectively). Displays neuroprotective effects in vivo and in vitro.

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Quantity 10 mg
Formula Weight 377.91
Percent Purity >99%
Chemical Name or Material Co 101244 hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'