missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNTN4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122085
This item is not returnable.
View return policy
Description
CNTN4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
Specifications
| CNTN4 | |
| Polyclonal | |
| Unconjugated | |
| CNTN4 | |
| 9630050B05; Axcam; axonal-associated cell adhesion molecule; Big-2; brain-derived immunoglobulin superfamily molecule; brain-derived immunoglobulin superfamily protein 2; CNTN4; CNTN4A; contactin 4; contactin-4; Kiaa3024; MGC33615; neural cell adhesion protein BIG-2; SCA16 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100153462, 152330 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8IWV2 | |
| CNTN4 | |
| Recombinant protein Contactin 4. The antigen corresponds to amino acid range 731-980 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction