missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNTN4 Polyclonal Antibody

Rabbit Polyclonal Antibody
£133.00 - £430.00
Specifications
| Antigen | CNTN4 |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17720464
![]() |
Invitrogen™
PA5119050 |
200 μL |
£430.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17733484
|
Invitrogen™
PA5122085 |
20 μL |
£133.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CNTN4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.Specifications
| CNTN4 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Pig | |
| CNTN4 | |
| 9630050B05; Axcam; axonal-associated cell adhesion molecule; Big-2; brain-derived immunoglobulin superfamily molecule; brain-derived immunoglobulin superfamily protein 2; CNTN4; CNTN4A; contactin 4; contactin-4; Kiaa3024; MGC33615; neural cell adhesion protein BIG-2; SCA16 | |
| CNTN4 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8IWV2 | |
| 100153462, 152330 | |
| Recombinant protein Contactin 4. The antigen corresponds to amino acid range 731-980 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title