missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOT1 Mouse anti-Human, Clone: 1A10A11, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CNOT1 |
|---|---|
| Clone | 1A10A11 |
| Concentration | 1 mg/mL |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16868445
|
Proteintech
66507-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16858445
|
Proteintech
66507-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This is a rabbit polyclonal antibody raised against a region from the C-terminal CNOT1 of human origin. CNOT1 exists as many isoforms. Isoform one is about 267 kDa and isoform four is about 173 kDa.
Scaffolding component of the CCR4-NOT complex which is one of the major cellular mRNA deadenylases and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation and general transcription regulation. Additional complex functions may be a consequence of its influence on mRNA expression. Its scaffolding function implies its interaction with the catalytic complex module and diverse RNA-binding proteins mediating the complex recruitment to selected mRNA 3′UTRs. Involved in degradation of AU-rich element (ARE)-containing mRNAs probably via association with ZFP36. Mediates the recruitment of the CCR4-NOT complex to miRNA targets and to the RISC complex via association with TNRC6A, TNRC6B or TNRC6C. Acts as a transcriptional repressor. Represses the ligand-dependent transcriptional activation by nuclear receptors. Involved in the maintece of emryonic stem (ES) cell identity.Specifications
| CNOT1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| AD 005, CCR4 associated factor 1, CDC39, CNOT1, hNOT1, KIAA1007, NOT1, NOT1H | |
| CNOT1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1A10A11 | |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| A5YKK6 | |
| 23019 | |
| CNOT1 Fusion Protein Ag5623 | |
| Primary | |
| -20°C | |
| CNOT1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title