missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOT1 Mouse anti-Human, Clone: 1A10A11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66507-1-IG-150UL
This item is not returnable.
View return policy
Description
This is a rabbit polyclonal antibody raised against a region from the C-terminal CNOT1 of human origin. CNOT1 exists as many isoforms. Isoform one is about 267 kDa and isoform four is about 173 kDa.
Scaffolding component of the CCR4-NOT complex which is one of the major cellular mRNA deadenylases and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation and general transcription regulation. Additional complex functions may be a consequence of its influence on mRNA expression. Its scaffolding function implies its interaction with the catalytic complex module and diverse RNA-binding proteins mediating the complex recruitment to selected mRNA 3′UTRs. Involved in degradation of AU-rich element (ARE)-containing mRNAs probably via association with ZFP36. Mediates the recruitment of the CCR4-NOT complex to miRNA targets and to the RISC complex via association with TNRC6A, TNRC6B or TNRC6C. Acts as a transcriptional repressor. Represses the ligand-dependent transcriptional activation by nuclear receptors. Involved in the maintece of emryonic stem (ES) cell identity.Specifications
| CNOT1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| A5YKK6 | |
| CNOT1 | |
| CNOT1 Fusion Protein Ag5623 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1A10A11 | |
| Unconjugated | |
| CNOT1 | |
| AD 005, CCR4 associated factor 1, CDC39, CNOT1, hNOT1, KIAA1007, NOT1, NOT1H | |
| Mouse | |
| Protein G | |
| RUO | |
| 23019 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction