missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CMG-2/ANTXR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-68911
This item is not returnable.
View return policy
Description
CMG-2/ANTXR2 Polyclonal specifically detects CMG-2/ANTXR2 in Mouse samples. It is validated for Western Blot.
Specifications
| CMG-2/ANTXR2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| anthrax toxin receptor 2, Capillary morphogenesis gene 2 protein, capillary morphogenesis protein 2, CMG2MGC111533, CMG-2MGC45856, FLJ31074, ISH, JHF | |
| Rabbit | |
| 53 kDa | |
| 100 μL | |
| Neuroscience | |
| 118429 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q6DFX2 | |
| ANTXR2 | |
| Synthetic peptides corresponding to Antxr2 (anthrax toxin receptor 2) The peptide sequence was selected from the N terminal of Antxr2. Peptide sequence GLVPSYAENEAKKSRSLGASVYCVGVLDFEQAQLERIADSKDQVFPVKGG. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction