missing translation for 'onlineSavingsMsg'
Learn More

Clusterin/APOJ Antibody (CLI-9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications

£146.00 - £443.00

Specifications

Antigen Clusterin/APOJ
Clone CLI-9
Concentration 1.0 mg/mL
Dilution Western Blot 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Applications Western Blot, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18084397
View Documents
Novus Biologicals
NBP1-97521-0.025MG
0.025 mg
£146.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18039276
View Documents
Novus Biologicals
NBP1-97521
0.1 mg
£443.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Clusterin/APOJ Monoclonal specifically detects Clusterin/APOJ in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Clusterin/APOJ
1.0 mg/mL
Western Blot, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence
Unconjugated
Mouse
Human
1191
Purified human clusterin (TRPM-2).
Primary
Store at 4C. Do not freeze.
CLI-9
Western Blot 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Monoclonal
Purified
RUO
40, 40, sulfated glycoprotein 2, Aging-associated gene 4 protein, aging-associated protein 4, apo-J, APOJ, Apolipoprotein J, CLIclusterin (complement lysis inhibitor, SP-40, clusterin, Complement cytolysis inhibitor, complement lysis inhibitor, Complement-associated protein SP-40, Ku70-binding protein 1, KUB1SGP2, MGC24903, NA1/NA2, SGP-2, SP-40, sulfated glycoprotein 2, Testosterone-repressed prostate message 2, testosterone-repressed prostate message 2, apolipoprotein J), TRPM-2TRPM2
CLU
IgG1
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.