missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clusterin/APOJ Antibody (CLI-9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-97521
This item is not returnable.
View return policy
Description
Clusterin/APOJ Monoclonal specifically detects Clusterin/APOJ in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Clusterin/APOJ | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| CLU | |
| Purified human clusterin (TRPM-2). | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| CLI-9 | |
| Unconjugated | |
| 40, 40, sulfated glycoprotein 2, Aging-associated gene 4 protein, aging-associated protein 4, apo-J, APOJ, Apolipoprotein J, CLIclusterin (complement lysis inhibitor, SP-40, clusterin, Complement cytolysis inhibitor, complement lysis inhibitor, Complement-associated protein SP-40, Ku70-binding protein 1, KUB1SGP2, MGC24903, NA1/NA2, SGP-2, SP-40, sulfated glycoprotein 2, Testosterone-repressed prostate message 2, testosterone-repressed prostate message 2, apolipoprotein J), TRPM-2TRPM2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 1191 | |
| Store at 4C. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction