missing translation for 'onlineSavingsMsg'
Learn More

Clotrimazole, Thermo Scientific™

Specific inhibitor of calcium-activated potassium channels

Brand:  Thermo Scientific Alfa Aesar J63895.06


Quantity 5g

 View more versions of this product

Product Code. 15486049

  • ONLINE EXCLUSIVE PRICE
    £60.60
    £54.65 / 5g
    Save £5.95
    9% Off
 Restricted Item
Estimated Shipment: 08-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Clotrimazole is an imidazole derivative and an antifungal compound and a CYP (cytochrome P450) inhibitor. Clotrimazole has been shown to block the intermediate-conductance, IK1 channels (Ca2+ activated K+ channels), in cells such as erythrocytes. In vitro studies of various yeast strains have demonstrated susceptibility to clotrimazole. Clotrimazole is an activator of MB67 and an inhibitor of CYP3A4 and CYP51A1.

Solubility
Soluble in chloroform, DMSO, DMF and alcohols

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

23593-75-1
344.842
VNFPBHJOKIVQEB-UHFFFAOYSA-N
2812
1-[(2-chlorophenyl)-diphenylmethyl]imidazole
C22H17ClN2
MFCD00057220
1-[(2-Chlorophenyl)diphenylmethyl]-1H-imidazole; 1-(o-Chlorotrityl)imidazole
CHEBI:3764
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3Cl)N4C=CN=C4

Specifications

23593-75-1
5g
MFCD00057220
1-[(2-Chlorophenyl)diphenylmethyl]-1H-imidazole; 1-(o-Chlorotrityl)imidazole
VNFPBHJOKIVQEB-UHFFFAOYSA-N
1-[(2-chlorophenyl)-diphenylmethyl]imidazole
2812
344.84
White
C22H17ClN2
14,2417
Soluble in chloroform, DMSO, DMF and alcohols
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3Cl)N4C=CN=C4
344.842
CHEBI:3764
Clotrimazole
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Clotrimazole

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contain: Clotrimazole

SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.