missing translation for 'onlineSavingsMsg'
Learn More

Clocinnamox mesylate, Tocris Bioscience™

Product Code. 18053782
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
This item is not returnable. View return policy

Product Code. 18053782

Brand: Tocris Bioscience 0898/2

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Irreversible μ antagonist

Systemically active, irreversible μ-opioid receptor antagonist (apparent Ki values are 0.7, 1.9 and 5.7 nM for mouse μ, δ and κ receptors respectively).
TRUSTED_SUSTAINABILITY

Specifications

CAS 117332-69-1
Molecular Formula C30H33ClN2O7S
Quantity 2 mg
Synonym clocinnamox mesylate, 14?-p-chlorocinnamoylamino-7,8-dihydro-n-cyclopropylmethylmorphinone mesylate
InChI Key XAXNKAGAUFXFDO-TYMYTQEGSA-N
SMILES CS(=O)(=O)O.C1CC1CN2CCC34C5C(=O)CCC3(C2CC6=C4C(=C(C=C6)O)O5)NC(=O)C=CC7=CC=C(C=C7)Cl
IUPAC Name (E)-N-[(4S,4aR,7aS,12bS)-3-(cyclopropylmethyl)-9-hydroxy-7-oxo-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-4a-yl]-3-(4-chlorophenyl)prop-2-enamide;methanesulfonic acid
Molecular Weight (g/mol) 601.111
PubChem CID 133655490
Formula Weight 601.11
Percent Purity >98%
Chemical Name or Material Clocinnamox mesylate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.