missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLN8 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£178.00 - £387.00
Specifications
| Antigen | CLN8 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:100 - 1:500, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18642231
|
Novus Biologicals
NBP2-92342-0.02ml |
0.02 mL |
£178.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18685270
|
Novus Biologicals
NBP2-92342-0.1ml |
0.1 mL |
£387.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLN8 Polyclonal antibody specifically detects CLN8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| CLN8 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 2055 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:100 - 1:500, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| C8orf61, ceroid-lipofuscinosis, neuronal 8 (epilepsy, progressive with mental retardation), EPMR | |
| A synthetic peptide corresponding to a sequence within amino acids 200-286 of human CLN8 (NP_061764.2). MFHCRMVLTYHMWWVCFWHWDGLVSSLYLPHLTLFLVGLALLTLIINPYWTHKKTQQLLNPVDWNFAQPEAKSRPEGNGQLLRKKRP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title