missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Human CLK1, GST Tag Recombinant Protein
£438.00 - £12060.00
Specifications
Accession Number | NP_004062 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 70.1 kDa |
Gene Alias | CLK1 |
KinaseFamily | CLK Kinase Family |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11543427
|
Thermo Scientific™
PV3315 |
10μg |
£438.00
10µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10185532
|
Thermo Scientific™
PV3984 |
1 mg |
£12060.00
1mg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CLK1 is a dual specificity kinase that phosphorylates PTP-1B and members of the SR family of splicing factors. It is thought to be involved in neuronal differentiation and mRNA splicing.Specifications
NP_004062 | |
70.1 kDa | |
CLK Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.05mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Kinases & Inhibitors | |
Liquid |
Kinase Assay | |
CLK1 | |
Human | |
GST Tag | |
Dry Ice | |
Recombinant, Catalytic |