missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CLK1, GST Tag Recombinant Protein

Product Code. 11543427
Change view
Click to view available options
Quantity:
1 mg
10 μg
Unit Size:
10µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11543427 10 μg 10µg
10185532 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11543427 Supplier Invitrogen™ Supplier No. PV3315

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Catalytic.

CLK1 is a dual specificity kinase that phosphorylates PTP-1B and members of the SR family of splicing factors. It is thought to be involved in neuronal differentiation and mRNA splicing. Therefore, the encoded protein may play an indirect role in governing splice site selection.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49759
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 0.05mM EDTA, 0.05% Triton X-100, 50% glycerol, 2mM DTT, 150mM NaCl and no preservative; pH 7.5
Gene ID (Entrez) 1195
Molecular Weight (g/mol) 70.1 kDa
Name Human CLK1, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias CDC like kinase 1; CDC28/CDC2-like kinase; CDC-like kinase 1; CLK; CLK/STY; CLK1; Dual specificity protein kinase CLK1; Protein kinase CLK1; protein kinase STY; protein tyrosine kinase STY; Sty
Common Name CLK1
Gene Symbol CLK1
KinaseFamily CLK Kinase Family
Protein Length 129-484
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 129-484
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical binding/kinetic, Biochemical phospho-specific activity (no antibody)
Kinase Group Ser⁄Thr Kinases: CMCG Group
Shipping Condition Dry Ice
System Compatibility Z'-LYTE™
Target Entry CLK1
Target Gene CDC-like kinase 1
Expression System E. coli
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Catalytic
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.