missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLIP170 Antibody (SR1120), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | CLIP170 |
|---|---|
| Clone | SR1120 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18672634
|
Novus Biologicals
NBP3-22556-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611964
|
Novus Biologicals
NBP3-22556-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLIP170 Monoclonal antibody specifically detects CLIP170 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| CLIP170 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CAP-GLY domain containing linker protein 1, CLIP, CLIP170, CLIP-170Reed-Sternberg intermediate filament-associated protein, CYLN1MGC131604, cytoplasmic linker 1, Cytoplasmic linker protein 1, Cytoplasmic linker protein 170 alpha-2, Reed-Steinberg cell-expressed intermediate filament-associated protein, Restin, restin (Reed-Steinberg cell-expressed intermediate filament-associated protein), RSNCAP-Gly domain-containing linker protein 1 | |
| A synthesized peptide derived from human CLIP170 (Uniprot #: P30622) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1120 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Dendritic Cell Markers | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 6249 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title