missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CLIP170 Monoclonal antibody specifically detects CLIP170 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
Specifications
| Antigen | CLIP170 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | SR1120 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Formulation | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Gene Alias | CAP-GLY domain containing linker protein 1, CLIP, CLIP170, CLIP-170Reed-Sternberg intermediate filament-associated protein, CYLN1MGC131604, cytoplasmic linker 1, Cytoplasmic linker protein 1, Cytoplasmic linker protein 170 alpha-2, Reed-Steinberg cell-expressed intermediate filament-associated protein, Restin, restin (Reed-Steinberg cell-expressed intermediate filament-associated protein), RSNCAP-Gly domain-containing linker protein 1 |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from human CLIP170 (Uniprot #: P30622) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?