missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Click-IT™ Alexa Fluor™ 647 DIBO Alkyne, for copper free click chemistry detection of azide

Product Code. 10538950
Click to view available options
Quantity:
500 μg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10538950

Brand: Molecular Probes™ C10408

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Click-IT™ Alexa Fluor™ 647 DIBO Alkyne, for copper free click chemistry detection of azide

The fluorescent Alexa Fluor™ 647 DIBO alkyne is reactive with azides via a copper-free click chemistry reaction.

Please consider this copper-free variation to our copper requiring alkynes if you are staining the surface of live cells or have concerns about native protein function loss with copper in cell extracts. Copper can damage fluorescent proteins, Quantum Dot nanocyrstals, certain enzymes and photoproteins like RPE. The DIBO reagent is not suitable for staining intracellular components of fixed and permeabilized cells due to high backgrounds.
TRUSTED_SUSTAINABILITY

Specifications

Description Click-IT™ Alexa Fluor™ 647 DIBO Alkyne, 500μg, Red/Blue, Solid, 668nm Emission Wavelength, 647nm Excitation Wavelength, ∽1500 Molecular Weight, Soluble in DMSO (Dimethylsulfoxide), Used for Labeling Chemistry Applications
Formulation Solid
For Use With (Application) Labeling chemistry
Quantity 500 μg
Product Type Click-IT™ Alexa Fluor™ 647 DIBO Alkyne
Molecular Weight (g/mol) ∽1500kDa
Content And Storage ≤-20°C
Label or Dye Alexa Fluor™ 488, Alexa Fluor™ 647
Color Red, Blue
Emission 668
Excitation 647
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.