missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Click-IT™ Alexa Fluor™ 488 DIBO Alkyne, for copper free click chemistry detection of azide
Click to view available options
Quantity:
500 μg
Unit Size:
Each
Description
The fluorescent Alexa Fluor™ 488 DIBO alkyne is reactive with azides via a copper-free click chemistry reaction.
Please consider this copper-free variation to our copper requiring alkynes if you are staining the surface of live cells or have concerns about native protein function loss with copper in cell extracts. Copper can damage fluorescent proteins, Quantum Dot nanocyrstals, certain enzymes and photoproteins like RPE. The DIBO reagent is not suitable for staining intracellular components of fixed and permeabilized cells due to high backgrounds.
Please consider this copper-free variation to our copper requiring alkynes if you are staining the surface of live cells or have concerns about native protein function loss with copper in cell extracts. Copper can damage fluorescent proteins, Quantum Dot nanocyrstals, certain enzymes and photoproteins like RPE. The DIBO reagent is not suitable for staining intracellular components of fixed and permeabilized cells due to high backgrounds.
Specifications
Specifications
| Description | Click-IT™ Alexa Fluor™ 488 DIBO Alkyne, 500μg, Green/Orange, Solid, 494nm Emission Wavelength, 488nm Excitation Wavelength, 1039.23 Molecular Weight, C53H62N6O12S2 Molecular Formula, Soluble in DMSO (Dimethylsulfoxide) |
| Formulation | Solid |
| For Use With (Application) | Labeling chemistry |
| Quantity | 500 μg |
| Product Type | Click-IT™ Alexa Fluor™ 488 DIBO Alkyne |
| Molecular Weight (g/mol) | 1039.23kDa |
| Content And Storage | ≤-20°C |
| Label or Dye | Alexa Fluor™ 488 |
| Color | Orange-Green |
| Emission | 494 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction