missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLGN Recombinant Rabbit Monoclonal Antibody (23GB3280), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | CLGN |
|---|---|
| Clone | 23GB3280 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30088736
|
Invitrogen™
MA553410 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088431
|
Invitrogen™
MA553411 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLGN Recombinant Monoclonal antibody specifically detects CLGN in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| CLGN | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O14967 | |
| 1047 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3280 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 4930459O04Rik; A2/6; AI528775; Calmegin; calnexin-T; Clgn; Cln; hypothetical protein LOC504256; LOW QUALITY PROTEIN: calmegin; MEG 1 antigen; Meg1; testis tissue sperm-binding protein Li 79P | |
| A synthetic peptide of human Calmegin (180-350AA). | |
| Primary | |
| -20°C | |
| CLGN |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title