missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLGN Recombinant Rabbit Monoclonal Antibody (23GB3280), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553410
This item is not returnable.
View return policy
Description
Calmegin is a testis-specific endoplasmic reticulum chaperone protein. CLGN may play a role in spermatogeneisis and infertility.Calmegin is a testis-specific endoplasmic reticulum chaperone protein. CLGN may play a role in spermatogeneisis and infertility.
Specifications
| CLGN | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CLGN | |
| 4930459O04Rik; A2/6; AI528775; Calmegin; calnexin-T; Clgn; Cln; hypothetical protein LOC504256; LOW QUALITY PROTEIN: calmegin; MEG 1 antigen; Meg1; testis tissue sperm-binding protein Li 79P | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1047 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3280 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14967 | |
| CLGN | |
| A synthetic peptide of human Calmegin (180-350AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction