missing translation for 'onlineSavingsMsg'
Learn More

Cletoquine Oxalate, TRC

Product Code. 30363733
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363733 5 mg 5mg
30357361 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30363733 Supplier Toronto Research Chemicals Supplier No. TRCC5735005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14142-64-4
Molecular Formula C16 H22 Cl N3 O . C2 H2 O4
Molecular Weight (g/mol) 397.85
Synonym Ethanol, 2-[[4-[(7-chloro-4-quinolinyl)amino]pentyl]amino]-, ethanedioate (1:1), Ethanol, 2-[[4-[(7-chloro-4-quinolyl)amino]pentyl]amino]-, oxalate (1:2) (8CI), Cletoquine Oxalate, Desethylhydroxychloroquine oxalate
IUPAC Name 2-[4-[(7-chloroquinolin-4-yl)amino]pentylamino]ethanol;oxalic acid
SMILES CC(CCCNCCO)Nc1ccnc2cc(Cl)ccc12.OC(=O)C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C16H22ClN3O.C2H2O4/c1-12(3-2-7-18-9-10-21)20-15-6-8-19-16-11-13(17)4-5-14(15)16;3-1(4)2(5)6/h4-6,8,11-12,18,21H,2-3,7,9-10H2,1H3,(H,19,20);(H,3,4)(H,5,6)
Formula Weight 397.1404
Percent Purity >95%
Chemical Name or Material Cletoquine Oxalate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.