CAS RN 14142-64-4
CAS RN 14142-64-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cletoquine Oxalate, TRC
CAS: 14142-64-4 Molecular Formula: C16 H22 Cl N3 O . C2 H2 O4 Molecular Weight (g/mol): 397.85 Synonym: Ethanol, 2-[[4-[(7-chloro-4-quinolinyl)amino]pentyl]amino]-, ethanedioate (1:1),Ethanol, 2-[[4-[(7-chloro-4-quinolyl)amino]pentyl]amino]-, oxalate (1:2) (8CI),Cletoquine Oxalate,Desethylhydroxychloroquine oxalate IUPAC Name: 2-[4-[(7-chloroquinolin-4-yl)amino]pentylamino]ethanol;oxalic acid SMILES: CC(CCCNCCO)Nc1ccnc2cc(Cl)ccc12.OC(=O)C(=O)O