missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC9a Rat anti-Mouse, Clone: 1F6, Novus Biologicals™
Rat Monoclonal Antibody
£403.00
Specifications
| Antigen | CLEC9a |
|---|---|
| Clone | 1F6 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL |
| Applications | Flow Cytometry, ELISA, Immunofluorescence |
Description
CLEC9a Monoclonal specifically detects CLEC9a in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
| CLEC9a | |
| 0.2 mg/mL | |
| Flow Cytometry, ELISA, Immunofluorescence | |
| Unconjugated | |
| Rat | |
| Mouse | |
| C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
| RBL-2H3 cells expressing Mouse CLEC9a fused to an HA epitope. | |
| Primary | |
| Store at 4°C. |
| 1F6 | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 283420 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title