missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC9a Rat anti-Mouse, Clone: 1F6, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP3-07516-100ug
This item is not returnable.
View return policy
Description
CLEC9a Monoclonal specifically detects CLEC9a in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| CLEC9a | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL | |
| C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
| RBL-2H3 cells expressing Mouse CLEC9a fused to an HA epitope. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, ELISA, Immunofluorescence | |
| 1F6 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 283420 | |
| Store at 4°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction