missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC4E Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£167.00 - £302.00
Specifications
| Antigen | CLEC4E |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2-4ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18157643
|
Novus Biologicals
NBP2-27093SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18183275
|
Novus Biologicals
NBP2-27093 |
0.1 mg |
£302.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLEC4E Polyclonal specifically detects CLEC4E in Human, Primate samples. It is validated for Western Blot.Specifications
| CLEC4E | |
| Western Blot 2-4ug/ml | |
| Polyclonal | |
| Rabbit | |
| Immunology | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| CLECSF9, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 9, C-type lectin domain family 4 member E, C-type lectin domain family 4, member E, Macrophage-inducible C-type lectin, mincle, MINCLEC-type lectin superfamily member 9 | |
| CLEC4E | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human, common chimpanzee, orangutan, and white-cheeked gibbon, 94% in crab-eating macaque, rhesus monkey, and sheep, 88% in cow, rat, and new world monkey, 82% in panda, dog, horse, and african bush elephant, 76% in guinea pig, and naked mole rat, 70% in mouse, and rabbit, 64% in Chinese hamster, opossum, and platypus. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Primate | |
| Q9ULY5 | |
| 26253 | |
| This antibody was raised against CLEC4E | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title