missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC4E Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27093
This item is not returnable.
View return policy
Description
CLEC4E Polyclonal specifically detects CLEC4E in Human, Primate samples. It is validated for Western Blot.
Specifications
| CLEC4E | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| CLECSF9, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 9, C-type lectin domain family 4 member E, C-type lectin domain family 4, member E, Macrophage-inducible C-type lectin, mincle, MINCLEC-type lectin superfamily member 9 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human, common chimpanzee, orangutan, and white-cheeked gibbon, 94% in crab-eating macaque, rhesus monkey, and sheep, 88% in cow, rat, and new world monkey, 82% in panda, dog, horse, and african bush elephant, 76% in guinea pig, and naked mole rat, 70% in mouse, and rabbit, 64% in Chinese hamster, opossum, and platypus. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 2-4ug/ml | |
| Q9ULY5 | |
| CLEC4E | |
| This antibody was raised against CLEC4E | |
| 0.1 mg | |
| Immunology | |
| 26253 | |
| Human, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction