missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC10A/CD301 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CLEC10A/CD301 |
|---|---|
| Concentration | 0.18 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16821884
|
Proteintech
13590-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16811884
|
Proteintech
13590-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may function as a cell surface antigen. Two transcript variants encoding distinct isoforms have been identified for this gene.Specifications
| CLEC10A/CD301 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q8IUN9 | |
| 10462 | |
| CLEC10A/CD301 Fusion Protein Ag4490 | |
| Primary | |
| -20°C | |
| CLEC10A |
| 0.18 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CD301, CLEC10A, CLECSF13, CLECSF14, HML, HML2, Macrophage lectin 2 | |
| CLEC10A | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title