missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC10A/CD301 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13590-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may function as a cell surface antigen. Two transcript variants encoding distinct isoforms have been identified for this gene.Specifications
| CLEC10A/CD301 | |
| Polyclonal | |
| Unconjugated | |
| CLEC10A | |
| CD301, CLEC10A, CLECSF13, CLECSF14, HML, HML2, Macrophage lectin 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10462 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8IUN9 | |
| CLEC10A | |
| CLEC10A/CD301 Fusion Protein Ag4490 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction