Learn More
Diversey™ ClearKlens Tego 2000 Disinfectant
Cleans and disinfects in sterile environments. The Diversey ClearKlens Tego 2000 Disinfectant is an active amphoteric disinfectant with a broad spectrum of activity. This microbiocidal disinfectant can be applied by means of a mop or cloth.
Brand: Diversey™ 7513387
Code : Z4
Additional Details : CAS Number : 139734-65-9 Weight : 2.50000kg Transport : UN number : 3082 Chem class : 9 Pack group : III
Description
- Hard surface disinfectant based on amphoretics
- Supplied with certificates of analysis, sterility, and irradiation where applicable
Specifications
∼8 (undiluted) | |
SC VH25 | |
50 mL |
Sterile | |
50 Pk. |
Safety and Handling
- Clearklens Tego 2000 (SC) VH28
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.