missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Screw-Top Bottle, Square, Clear, Soda-Lime Glass, Narrow Mouth, High form DIN thread

Product Code. 11716928 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11716928 1000 mL Pack of 10
12850924 100 mL Pack of 10
11776918 250 mL Pack of 10
11706928 500 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11716928 Supplier DWK Life Sciences Supplier No. 238105453

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Square clear soda lime glass bottles, narrow mouth.

Square clear soda lime (Type 3) glass bottles, narrow mouth, high form DIN thread.

  • Square shape for efficient packing
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
For Use With (Application) Storage of samples and reagents; Sample collection; Transport and shipping
Material Soda Lime Glass
Quantity 10/Pk.
Thread Size (Metric) GL 45
Diameter (Metric) 97 mm
Autoclavable Not Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.