missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Screw-Top Bottle, Square, Clear, Soda-Lime Glass, Narrow Mouth, High form DIN thread
Shop All DWK Life Sciences ProductsDescription
Square clear soda lime (Type 3) glass bottles, narrow mouth, high form DIN thread.
- Square shape for efficient packing
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| For Use With (Application) | Storage of samples and reagents; Sample collection; Transport and shipping |
| Material | Soda Lime Glass |
| Quantity | 10/Pk. |
| Thread Size (Metric) | GL 32 |
| Diameter (Metric) | 77 mm |
| Autoclavable | Not Autoclavable |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?