Learn More
Description
The classical stoppered reagent bottles are moulded from DURAN™ borosilicate 3.3 Type 1 glass, and are suitable for the storage of liquid and solid chemicals and reagents. The bottles feature a narrow conical ground joint neck and comply with ISO 4796-2. The narrow neck is suitable for liquids and the angle of the shoulders ensures that the contents can be smoothly poured. The glass stopper provides an excellent seal that protects the contents from evaporation, oxidation, and contamination. The bottles have a Retrace Code for manufacturing lot traceability and a marking field in permanent white enamel.
- All borosilicate glass components and therefore suitable for storage of aggressive materials
- Tight closure due to standard ground joint
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic print
Specifications
Specifications
| Capacity (Metric) | 5000 mL |
| Description | Neck with standard ground joint and glass flat-headed stopper |
| For Use With (Application) | Storage of air sensitive reagents, pharmaceuticals, and chemicals; Storage of hygroscopic liquids and pharmaceuticals; Transport and shipping of samples; Storage of hygroscopic liquids and pharmaceuticals |
| Material | Borosilicate Glass (3.3) |
| Quantity | 1 Ea. |
| Diameter (Metric) | 182 mm |
| Autoclavable | Autoclavable |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.