Learn More
Fisherbrand™ Clear Graduated Glass Bottles with Black Phenolic Caps
GREENER_CHOICE
Product Code. p-7145765 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
60 mL
240 mL
Unit Size:
Pack of 48
Product Code. 16134387
Only null left
Add to basket
Add to basket
Fisherbrand™ Clear Graduated Glass Bottles with Black Phenolic Caps feature a wide mouth for easy removal of contents.
  • Graduated in both milliliters and ounces
  • Clear glass bottles are designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents
Specifications
Capacity (Metric) 60 mL
Capacity (English) 2 oz.
Color Clear
Material Glass (Type 3)
Quantity 48
Closure Color Black
Neck Finish 38-400
Closure Included Yes
Graduated Yes
Mouth Wide
Closure Liner Polyethylene Cone
Closure Material Phenolic