missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Graduated Glass Bottle Beakers with White Polypropylene Pulp/Vinyl Cap
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
30 mL
120 mL
Neck Finish:
33-400
48-400
Quantity:
144/Cs.
432/Cs.
Unit Size:
Pack of 144
Pack of 432
Description
Includes
Polypropylene Pulp/Vinyl lined cap
Graduated Medium Rounds are excellent multipurpose containers for use with biological and pathological specimens, but can also be used for storing industrial laboratory chemicals and reagents. These clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling. The wide mouth facilitates easy mixing, handling, and transfer of contents. Graduated in both milliliters and ounces.
- Graduated in both milliliters and ounces
- Clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling
- Wide mouth facilitates easy mixing, handling, and transfer of contents
White Polypropylene Pulp/Vinyl Lined Caps
- Polypropylene Caps are known for good impact strength, durability, cost effectiveness, thermal resistance, and pliability
- Caps are autoclavable
- Liner is made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
Specifications
Specifications
| Capacity (Metric) | 30 mL |
| Capacity (English) | 1 oz. |
| Color | Clear |
| Material | Glass |
| Quantity | 432/Cs. |
| Closure Color | White |
| Liner Material | Pulp/Vinyl |
| Neck Finish | 33-400 |
| Certifications/Compliance | USP Type III |
| Closure Included | Yes |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction