missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Graduated Glass Bottle Beakers with White Polypropylene PE Foam/PTFE Cap

Product Code. 15875472 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
120 mL
Neck Finish:
48-400
Quantity:
24/Cs.
Unit Size:
Pack of 24
This item is not returnable. View return policy

Product Code. 15875472

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Polypropylene PE Foam/PTFE lined cap

Environmentally sensitive bottles help eliminate waste. Fisherbrand™ Clear Graduated Clear Graduated Boston Round Bottles with White Polypropylene PE Foam/PTFE Lined Cap ensures product integrity for long term storage.

Graduated Medium Rounds are excellent multipurpose containers for use with biological and pathological specimens, but can also be used for storing industrial laboratory chemicals and reagents. These clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling. The wide mouth facilitates easy mixing, handling, and transfer of contents. Graduated in both milliliters and ounces.

Clear Graduated Boston Round Bottles

  • Feature a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Clear (flint) glass is appropriate when visibility is important but protection from UV rays is not necessary

Polypropylene PE Foam/PTFE Lined Caps

  • Offer good impact strength, thermal resistance and pliability
  • Autoclavable
  • Liners consist of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
  • Liners are very resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference

Specifications

Capacity (Metric) 120 mL
Capacity (English) 4 oz.
Color Clear
Material Glass
Quantity 24/Cs.
Closure Color White
Liner Material PE Foam/PTFE
Neck Finish 48-400
Closure Included Yes
Graduated Yes
Mouth Wide
Shape Round
Product Type Graduated Medium Round Bottle
Closure Liner With Liner
Closure Material Polypropylene
Closure Size 48-400 mm
Diameter (Metric) 51 mm
Closure Type Screw Cap
Through the Closure Dispensing Yes
Autoclavable Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.