missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLCN5 Rabbit anti-Canine, Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CLCN5 |
|---|---|
| Concentration | 0.4 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16865165
|
Proteintech
26812-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16855165
|
Proteintech
26812-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Proton-coupled chloride transporter. Functions as antiport system and exchanges chloride ions against protons. Important for normal acidification of the endosome lumen. May play an important role in renal tubular function.Specifications
| CLCN5 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Canine, Human, Mouse | |
| P51795, Q9WVD4 | |
| 1184, 12728, 491882 | |
| CLCN5 Fusion Protein Ag25182 | |
| Primary | |
| -20°C | |
| CLCN5 |
| 0.4 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| chloride channel 5, Chloride channel protein 5, Chloride transporter ClC 5, ClC 5, CLC5, CLCK2, CLCN5, DENTS, hCIC K2, hClC K2, XLRH, XRN | |
| CLCN5 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title