missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLCN5 Rabbit anti-Canine, Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26812-1-AP-20UL
This item is not returnable.
View return policy
Description
Proton-coupled chloride transporter. Functions as antiport system and exchanges chloride ions against protons. Important for normal acidification of the endosome lumen. May play an important role in renal tubular function.Specifications
| CLCN5 | |
| Polyclonal | |
| Unconjugated | |
| CLCN5 | |
| chloride channel 5, Chloride channel protein 5, Chloride transporter ClC 5, ClC 5, CLC5, CLCK2, CLCN5, DENTS, hCIC K2, hClC K2, XLRH, XRN | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1184, 12728, 491882 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P51795, Q9WVD4 | |
| CLCN5 | |
| CLCN5 Fusion Protein Ag25182 | |
| 20 μL | |
| Primary | |
| Canine, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction