missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLCN1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69124
This item is not returnable.
View return policy
Description
CLCN1 Polyclonal specifically detects CLCN1 in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| CLCN1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CLCN1 | |
| Synthetic peptides corresponding to Clcn1 (chloride channel 1) The peptide sequence was selected from the C terminal of Clcn1. Peptide sequence SSIFQRLLHCLLGKAHSTKKKITQDSTDLVDNMSPEEIEAWEREQLSQPV. | |
| Affinity purified | |
| RUO | |
| 1180 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry | |
| chloride channel 1, skeletal muscle, Chloride channel protein, skeletal muscle, clC-1, CLC1chloride channel protein 1, MGC138361, MGC142055 | |
| Rabbit | |
| 110 kDa | |
| 100 μL | |
| Primary | |
| Pig: 86%. | |
| Mouse, Rat, Bovine, Canine, Equine, Equine, Guinea Pig, Goat, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction