missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Claudin 11 Recombinant Rabbit Monoclonal Antibody (23GB5090), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | Claudin 11 |
|---|---|
| Clone | 23GB5090 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088473
![]() |
Invitrogen™
MA552786 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088772
|
Invitrogen™
MA552787 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Claudin 11 Recombinant Monoclonal antibody specifically detects Claudin 11 in Human samples. It is validated for Western BlotSpecifications
| Claudin 11 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O75508 | |
| 5010 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5090 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| claudin 11; claudin 11 (oligodendrocyte transmembrane protein); Claudin11; claudin-11; Cldn11; hypothetical protein LOC508268; oligodendrocyte transmembrane protein; Oligodendrocyte-specific protein; OSP; OTM | |
| A synthesized peptide derived from human Oligodendrocyte Specific Protein (25-80AA). | |
| Primary | |
| -20°C | |
| Cldn11 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title